Learn More
Bulbman Inc FCR 64625 HLX Osram Photo Optic 54248 Light Bulb
SDP
Supplier:  Bulbman Inc 104400/54248
Encompass_Preferred
FCR 64625 HLX Osram Photo Optic 54248 Light Bulb: 12 volt, 100 watt, quartz halogen T3.5 lamp with a GY6.35 bipin base.
Catalog No. NC1703565