Learn More
Medchemexpress LLC FCRL2 Human HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7692450UG
Encompass_Preferred
The FCRL2 protein regulates normal and neoplastic B cell development and has been shown to be involved in complex processes that control B cell maturation Its tyrosine-phosphorylated isoform 2 interacts with PTPN6 suggesting the existence of a signaling pathway and emphasizing the role of this protein in cellular regulation FCRL2 Protein Human (HEK293 His) is the recombinant human-derived FCRL2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85722