Learn More
Apexbio Technology LLC DiscoveryProbe™ FDA-approved Drug Library,100uL/well(10 mM solution),96 Well Racks With Screw Cap
SDP
Supplier:  Apexbio Technology LLC L1021.11
Encompass_Preferred
DiscoveryProbe™ FDA-approved drug library includes 1971 FDA approved drugs for high throughput screening (HTS) and high content screening (HCS). It can be used to find new targets for old drugs. The bioactivity and safety of these drugs were confirmed by clinical trials. As they are all FDA-approved drugs, the drugs screened can be used for clinical testing directly. Other sizes are also available. Please inqury us for quote.Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0855
Add to Cart
Request a Quote