Learn More
Medchemexpress LLC FDI-6 | 313380-27-7 | 99.6% | 437.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11272150MG
Encompass_Preferred
FDI-6 is an inhibitor of FOXM1, designed for research use only. It directly binds to the FOXM1 protein, displacing it from genomic targets in MCF-7 breast cancer cells and leading to transcriptional down-regulation.
  • Inhibits FOXM1 protein
  • Displaces FOXM1 from genomic targets
  • Induces transcriptional down-regulation
  • Suitable for research applications
Catalog No. 50-004-37455