Learn More
Selleck Chemical LLC FDW028 E4732-25MG
SDP
Supplier:  Selleck Chemical LLC E473225MG
Encompass_Preferred
FDW028 is a highly potent and selective inhibitor of FUT8 FDW028 demonstrates strong anti-tumor effects by promoting defucosylation and inducing lysosomal degradation of B7-H3 via the chaperone-mediated autophagy (CMA) pathway Additionally FDW028 suppresses the AKT/mTOR signaling pathway FDW028 exhibits potent anti-Metastatic colorectal cancer (mCRC) activities
Catalog No. 50-000-41345