Learn More
SPEX CERTIPREP LLC 3P Iron at 1,000 ug/mL in 2% HNO3, 100 mL
Supplier:  SPEX CERTIPREP LLC 3P 1000261100
Encompass_Preferred
(HPS by Zeptometrix) Iron (Fe) ICP Single-Component Standard at 1,000 ug/mL in 2% HNO3.
Catalog No. NC1013786