Learn More
Electron Microscopy Sciences Feather Safeshield™ Blade #25
SDP
Supplier:  Electron Microscopy Sciences 7204325
Encompass_Preferred
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures. They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Catalog No. 50-334-94