Learn More
Medchemexpress LLC Febantel | 58306-30-2 | 99.9% | 446.48 | 1 G
SDP
Supplier:  Medchemexpress LLC HY175971G
Encompass_Preferred
Febantel is an oral dewormer used to treat gastrointestinal nematodes in sheep, as well as roundworm and tapeworm infections in poultry and livestock. It also inhibits the embryonic development of mouse hairworms and whipworm eggs.
  • Acts as an oral dewormer
  • Inhibits embryonic development of certain parasites
  • Suitable for research use only
Catalog No. 50-003-87136