Learn More
Medchemexpress LLC Febantel | 58306-30-2 | 99.9% | 446.48 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1759710MM/1ML
Encompass_Preferred
Febantel is an oral dewormer used to treat gastrointestinal nematodes in sheep, as well as roundworm and tapeworm infections in poultry and livestock. It also inhibits the embryonic development of mouse hairworms and whipworm eggs.
  • Oral dewormer
  • Treats gastrointestinal nematodes in sheep
  • Treats roundworm and tapeworm infections in poultry and livestock
  • Inhibits embryonic development of mouse hairworms and whipworm eggs
  • High purity: 99.91%
  • Available in solid and solution forms
Catalog No. 50-003-87135