Learn More
Medchemexpress LLC Febantel | 58306-30-2 | 99.9% | 446.48 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1759710G
Encompass_Preferred
Febantel is an oral dewormer utilized for treating gastrointestinal nematodes in sheep, as well as roundworm and tapeworm infections in poultry and livestock. It also hinders the embryonic development of mouse hairworms and whipworm eggs.
  • Treats gastrointestinal nematodes in sheep
  • Treats roundworm and tapeworm infections in poultry and livestock
  • Inhibits embryonic development of mouse hairworms
  • Inhibits embryonic development of whipworm eggs
Catalog No. 50-003-87454