Learn More
Selleck Chemical LLC Febantel S5092-25mg
SDP
Supplier:  Selleck Chemical LLC S509225MG
Encompass_Preferred
Febantel (Rintal Combotel Oratel Bay Vh 5757 Negabot Plus) is a prodrug that is metabolized to fenbendazole and oxfendazole which are undoubtedly the active parasiticides
Catalog No. 50-311-2237