Learn More
Medchemexpress LLC Febrifugine dihydrochloride | 32434-42-7 | 5 MG
Supplier: Medchemexpress LLC HYN2384A5MG
Febrifugine dihydrochloride is a quinazolinone alkaloid with demonstrated antimalarial activity against P. falciparum. It also exhibits anti-cancer properties by inhibiting DNA synthesis, inducing apoptosis, and reducing steroids in bladder cancer cells.
- Quinazolinone alkaloid with antimalarial activity
- Inhibits P. falciparum
- Demonstrates anti-cancer properties
- Inhibits DNA synthesis
- Induces apoptosis
- Reduces steroids in bladder cancer cells
- Increases eIF2α phosphorylation
- Inhibits asexual blood stage of P. falciparum 3D7 parasite strain
- Inhibits proliferation growth of bladder cancer cells
- Increases cells in G1 phase and decreases cells in G2 phase
- Reduces mortality and parasitemia in Plasmodium berghei infected mice
- Inhibits bladder cancer xenograft proliferation in nude mice