Learn More
Medchemexpress LLC Febrifugine dihydrochloride | 32434-42-7 | MFCD27978373 | 100.0% | 374.26 g/mol | C16H21Cl2N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN2384A10MG
Encompass_Preferred
Febrifugine dihydrochloride is a quinazolinone alkaloid research compound supplied as the dihydrochloride salt. It exhibits antimalarial activity by inhibiting Plasmodium falciparum and has reported antiproliferative effects in bladder cancer models; provided as a high-purity solid for in vitro and in vivo research.
  • Antimalarial activity against Plasmodium falciparum.
  • Mechanism involves increased eIF2α phosphorylation leading to parasite inhibition.
  • High purity as provided by manufacturer (99.95%).
  • White to off-white solid formulation, stable when stored frozen and protected from moisture and light.
  • Supplied in small research pack sizes suitable for in vitro and in vivo studies, including 10 mg packs.
Catalog No. 50-002-89600