Learn More
Cayman Chemical FebrIfugInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3704410 MG
Encompass_Preferred
A quinazolinone with antimalarial activity; reduces parasitemia and decreases mortality in mice infected with the P. berghei strain NK65 at 1 mg/kg per day
Catalog No. 50-291-4918