Learn More
Selleck Chemical LLC Febuxostat S1547-1g
SDP
Supplier:  Selleck Chemical LLC S15471G
Encompass_Preferred
Febuxostat (TMX 67 TEI-6720) is a selective xanthine oxidase inhibitor with Ki of 0 6 nM
Catalog No. 50-311-1135