Learn More
Medchemexpress LLC Febuxostat-d7 | 1285539-74-3 | 99.1% | C16H9D7N2O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14268S110MG
Encompass_Preferred
Febuxostat-d7 is a deuterium-labeled analogue of febuxostat used as a stable-isotope internal standard for analytical and pharmacokinetic studies. It preserves the febuxostat scaffold and enables mass-differentiated tracer measurements by LC-MS for accurate quantitation.
  • Deuterium labeled internal standard for analytical assays.
  • Molecular formula C16H9D7N2O3S and molecular weight 323.42.
  • High purity (99.06% by HPLC) suitable for quantitative analysis.
  • White to off-white solid; store powder at -20°C and solutions at -80°C for long-term stability.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-61872