Learn More
Medchemexpress LLC Febuxostat-d9 | 1246819-50-0 | MFCD21363615 | 98.7% | C16H7D9N2O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14268S10MG
Encompass_Preferred
Febuxostat-d9 is a deuterium-labeled analogue of febuxostat supplied as an isotopically labeled reference standard for analytical and research applications. It is intended for use as an internal standard in LC- or GC-MS assays and in studies of xanthine oxidase inhibition. The material is supplied with high isotopic enrichment and high chemical purity (≈98.7%, C16H7D9N2O3S, MW 325.43) for research use only.
  • Deuterium-labeled internal standard for LC- or GC-MS quantification.
  • Facilitates accurate quantification and compensation for matrix effects.
  • High isotopic enrichment and purity for reliable analytical performance.
  • Suitable for pharmacokinetic, metabolic, and bioanalytical studies.
  • Provides a clear mass shift from the unlabeled analyte for MS separation.
Catalog No. 50-002-59671