Learn More
Medchemexpress LLC Febuxostat impurity 7 | 1350352-70-3 | MFCD31807379 | 98.2% | 334.39 g/mol | C16H18N2O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1312695MG
Encompass_Preferred
Febuxostat impurity 7 is an analytical reference standard representing a synthetic impurity of febuxostat (CAS 1350352-70-3). It is intended for impurity identification, method development, and quality control applications in pharmaceutical analysis. The compound has molecular formula C16H18N2O4S and a molecular weight of 334.39 g/mol.
  • High purity suitable for analytical use (≈98.2%).
  • Molecular formula C16H18N2O4S and molecular weight 334.39 g/mol.
  • Supplied in milligram quantities for reference standard workflows.
  • Intended for impurity characterization and method development.
  • Provided with product data and certificate of analysis for traceability.
Catalog No. 50-002-64131