Learn More
Selleck Chemical LLC Febuxostat 10mg 144060-53-7 TMX 67, TEI-6720
SDP
Supplier:  Selleck Chemical LLC S154710MG
Encompass_Preferred
Febuxostat (TMX 67, TEI-6720) is a selective xanthine oxidase inhibitor with Ki of 0.6 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-770-3