Learn More
Medchemexpress LLC HY-14887 5mg Medchemexpress, Fedovapagon CAS:347887-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY148875MG
Encompass_Preferred
Medchemexpress, HY-14887 5mg Fedovapagon CAS:347887-36-9 Fedovapagon is a selective vasopressin V2 receptor (V2R) agonist with an EC 50 of 24 nM, which is being developed for the treatment of nocturia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2714