Learn More
Medchemexpress LLC Fedratinib (hydrochloride hydrate) | 1374744-69-0 | MFCD28137677 | 99.9% | 615.6 g/mol | C27H40Cl2N6O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10409A10MM 1ML
Encompass_Preferred
Fedratinib (hydrochloride hydrate) is the dihydrochloride monohydrate salt of fedratinib, a potent ATP-competitive JAK2 inhibitor used for research into JAK2 signaling and myeloproliferative disorders. Supplied as a ready-to-use solution, it supports biochemical and cell-based assays requiring a high-quality JAK2 inhibitor.
  • Potent JAK2 inhibitor with low-nanomolar potency
  • Dihydrochloride monohydrate salt form for improved solubility
  • Provided as a pre-made 10 mM solution for immediate use
  • High chemical purity (99.9%) for reliable experimental results
  • Suitable for biochemical and cell-based assay applications
Catalog No. 50-002-43541