Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fedratinib (hydrochloride hydrate) | 1374744-69-0 | MFCD28137677 | 99.9% | 615.62 g/mol | C27H40Cl2N6O4S | 2 G
SDP

Supplier:  Medchemexpress LLC HY10409A2G

Encompass_Preferred

Fedratinib hydrochloride hydrate is the dihydrochloride monohydrate salt of fedratinib, an ATP-competitive, orally active and selective JAK2 inhibitor supplied for research use. The compound is provided as a solid (white to yellow) with high reported purity and is intended for in vitro and preclinical studies; not for human therapeutic use.

  • High purity (≈99.9%) as reported by supplier.
  • Molecular weight 615.62 g/mol; formula C27H40Cl2N6O4S.
  • Packaged in small-scale quantities, commonly available as 2 G.
  • Recommended storage: solid at 4°C; in solution -20°C to -80°C.
  • Potent JAK2 inhibitor with reported IC50 ≈3 nM, suitable for signaling and oncology research.

Catalog No. 50-002-43486


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.