Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fedratinib hydrochloride hydrate | 1374744-69-0 | MFCD28137677 | 99.9% | 615.62 g·mol⁻¹ | C27H40Cl2N6O4S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY10409A5MG

Encompass_Preferred

Fedratinib hydrochloride hydrate is the hydrochloride hydrate salt of fedratinib, a selective, orally active, ATP-competitive JAK2 inhibitor used in preclinical research. The compound is supplied as a solid research reagent for studies of JAK2-driven signaling and cellular assays.

  • Hydrochloride hydrate salt form for increased stability
  • Selective ATP-competitive JAK2 inhibitor suitable for preclinical studies
  • Molecular weight 615.62 g·mol⁻¹ and formula C27H40Cl2N6O4S
  • High purity (≈99.9%) and solid, white to yellow appearance
  • Offered in small research pack sizes for dosing and assay work
  • Storage: 4°C sealed; in solvent, -80°C for long-term stability

Catalog No. 50-002-43210


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.