Learn More
Grainger Feed Through Stringer 6 Boxes 60 ft 20A
Supplier:  Grainger 14A296
Encompass_Preferred
The KH Industries Feed-Through Power Stringer is a heavy-duty power stringer designed for pulling electrical wires and cables through conduit. It features a powerful 1/2 HP motor with variable speed control that provides 0-150 feet per minute line speed. The stringer has a maximum pulling capacity of 1,000 lbs and can accommodate wire sizes from #4 to 500 MCM. It includes an adjustable brake system to prevent overruns and smooth wire pulling. The unit's compact size allows for easy maneuverability and portability on the jobsite. With its durable steel construction, this stringer provides reliable performance for electrical contractors pulling wires on commercial and industrial projects.
Catalog No. 50-275-0304