Learn More
Kuhner Shaker Inc FEEDBEADS GLUCOSE 12MM 40MG
Supplier:  Kuhner Shaker Inc SMFB02001
Encompass
FeedBeads- Glucose, 12 mm, 40 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. NC1671492