Learn More
ATCC Stem Cell Freezing Media (quantity of three)
Supplier:  ATCC ACS30203
Encompass
Volume: 20 mL Storage: 4°C Stem Cell Freezing Media is a serum­free, xeno­free, complete medium for the cryopreservation of various types of human and non-human animal stem cells. The ready-­to-­use solution is quick and convenient, requiring no additional supplements. This product has been qualified for use with primary tissue derived organoids, embryonic stem cells (hESC), induced pluripotent stem cells (hiPSC), and iPSCs-derived cells. Stem Cell Freezing Media can also be used with other continuous and cancer cell lines.
Catalog No. 50-238-1345