Learn More
TARGETMOL CHEMICALS INC Felbamate 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T194625MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Felbamate (W-554) is an Anti-epileptic Agent. The physiologic effect of felbamate is by means of Decreased Central Nervous System Disorganized Electrical Activity. Purity 99.44%
Catalog No. 50-287-8276