Learn More
Cayman Chemical Felbamate-d4 1mg
SDP
Supplier:  Cayman Chemical 284891 MG
Encompass_Preferred
An internal standard for the quantification of felbamate by GC- or LC-MS
Catalog No. 50-293-7799