Learn More
Vmrd Inc Feline Panleukopenia Virus (FPLV) FITC Conjugate Mab | 10mL
SDP
Supplier:  Vmrd Inc CJFFPLMAB10ML
Encompass
Anti-Feline Panleukopenia Virus (FPL) monoclonal antibody conjugated to fluorescein isothiocyanate. Murine origin. Suitable for staining FPL in tissues or cell cultures. Cross-reacts with mink enteritis virus.
Catalog No. NC1811104