Learn More
Kingfisher Biotech Inc Feline PD-L1 Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP2204F025
Encompass
The Feline PD-L1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline PD-L1 applications are for cell culture, ELISA standard, and Western Blot Control. Feline PD-L1 yeast-derived recombinant protein can be purchased in multiple sizes. Feline PD-L1 Specifications: (Molecular Weight: 24.6 kDa) (Amino Acid Sequence: FTITVSKDLY VVEYGSNVTM ECRFPVEEQL DLVSLIVYWE MEDKKIIQFV QGKEDLKVQH RSYSQRAQLL KDQLFLGKAA LQITNVTLED AGVYCCLIGY GGADYKRITL KVHAPYRKIN QRISVDPVTS EHELMCQAEG YPTAEVIWTN SAHQVLNGKT IISVSNMETK LFNVTSTLRI NTTANEIFYC TFLQRSSPEG NSTAELVIPE PFLVPANER (219)) (Gene ID: 100127110 (feline)). For research use only. Made in the USA
Catalog No. 50-296-4650