Learn More
Apexbio Technology LLC Felodipine 72509-76-3 50mg
SDP
Supplier:  Apexbio Technology LLC B141650
Encompass_Preferred
Felodipine (CAS 72509-76-3) is a dihydropyridine derivative that acts as a selective inhibitor of L-type calcium channels It binds selectively to L-type voltage-gated Ca channels and inhibits calcium influx displaying an IC50 of approximately 0 15 nM In biomedical research Felodipine is predominantly used to study calcium channel-mediated cellular processes including vascular smooth muscle contraction cardiovascular physiology and calcium signaling pathways
Catalog No. 50-000-62061