Learn More
Medchemexpress LLC Felodipine | 72509-76-3 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0309
Encompass_Preferred
Felodipine is a dihydropyridine, potent, and vasoselective calcium channel antagonist. It lowers blood pressure by selectively acting on vascular smooth muscle, especially in the resistance vessels. As an anti-hypertensive agent, Felodipine induces autophagy and can cross the blood-brain barrier.
  • Potent, vasoselective calcium channel antagonist
  • Lowers blood pressure
  • Induces autophagy
  • Can cross the blood-brain barrier
Catalog No. 50-004-50985