Learn More
Medchemexpress LLC Felypressin acetate | 914453-97-7 | 99.6% | 1040.2 g/mol | C46H65N13O11S2 · xC2H4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYA0182A10MG
Encompass_Preferred
Felypressin acetate is a synthetic non-catecholamine vasoconstrictor and a vasopressin V1 receptor agonist used as a research reagent and as a vasoconstrictive agent in dental applications. It is supplied as a high-purity peptide powder and is intended for laboratory use only.
  • Non-catecholamine vasoconstrictor and V1 receptor agonist.
  • White to off-white solid powder with high purity (99.6%).
  • Peptide sequence Cys-Phe-Phe-Gln-Asn-Cys-Pro-Lys-Gly-NH2 with 1-6 disulfide bridge.
  • Available as a 10 mg package for research use.
  • Store sealed, away from moisture; follow frozen storage recommendations for long-term stability.
Catalog No. 50-002-97899