Learn More
Medchemexpress LLC Felypressin acetate | 914453-97-7 | 99.6% | C46H65N13O11S2.xC2H4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0182A100MG
Encompass_Preferred
Felypressin acetate is a non-catecholamine vasoconstrictor and a vasopressin 1 agonist primarily used in dental procedures. Its vasoconstrictor action is mediated by V1 receptors of the blood vessel smooth muscle cells, contributing to reduced toxicity during dental procedures.
  • Non-catecholamine vasoconstrictor
  • Vasopressin 1 agonist
  • Widely used in dental procedures
  • Action mediated by V1 receptors
  • May reduce toxicity during dental procedures
  • May increase diastolic blood pressure in hypertensive patients
Catalog No. 50-004-33513