Learn More
Medchemexpress LLC Felypressin acetate | 914453-97-7 | MFCD16652603 | 99.6% | C46H65N13O11S2.xC2H4O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0182A50MG
Encompass_Preferred
Felypressin acetate is a non-catecholamine vasoconstrictor and a vasopressin 1 agonist. It is widely used in dental procedures.
  • Non-catecholamine vasoconstrictor: It constricts blood vessels without containing catecholamine.
  • Vasopressin 1 agonist: It activates vasopressin 1 receptors.
  • Dental procedures: It is widely used in dentistry, and can reduce toxicity during these procedures.
  • Mediated by V1 receptors: Its vasoconstrictor action is mediated by V1 receptors of blood vessel smooth muscle cells.
  • Increases diastolic blood pressure: It can increase the diastolic blood pressure of hypertensive patients with controlled blood pressure.
Catalog No. 50-004-32901