Learn More
Medchemexpress LLC Felypressin acetate (PLV-2 acetate) | 914453-97-7 | 99.60% | C46H65N13O11S2.xC2H4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0182A5MG
Encompass_Preferred
Felypressin acetate (PLV-2 acetate) is a non-catecholamine vasoconstrictor and a vasopressin 1 agonist. It is widely used in dental procedures. Its vasoconstrictor action is believed to be mediated by V1 receptors in the smooth muscle cells of blood vessels.
  • Acts as a non-catecholamine vasoconstrictor
  • Functions as a vasopressin 1 agonist
  • Widely used in dental procedures
  • Helps reduce toxicity during dental procedures
  • Can increase diastolic blood pressure in hypertensive patients with controlled blood pressure
  • Synthetic hormone with vasoconstrictor properties
  • Mediates vasoconstrictor action via V1 receptors
Catalog No. 50-004-32883