Learn More
COLDER PRODUCTS COMPANY FEMALE COUPLING 1/8 25/PK
Supplier:  COLDER PRODUCTS COMPANY NS4D17002
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 53 mm 2 09 in Gray with Dark Gray Overmold For Use With 3 2mm Includes Shut-off valves Polypropylene Hose Barb Valved In-line Coupling Body Height 1 05 in 26 6 mm
Catalog No. 50-313-1423