Learn More
AVANTOR MASTERFLEX (VWR LLC) Female luer x 3/32"" hose barb adapter, Nylon, 25/pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550202
Encompass
Female Luer x 3/32"ID Hose Barb Adapter, Nylon, 25/Pk, offers a variety of barbed luer adapters to meet your application's needsStandard hose barb for use with flexible tubingSmooth bore hose barb to reduce flow turbulence and risk of line contaminationWide-bore hose barb for enhanced fluid path flowLow-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Catalog No. NC1040149