Learn More
Medchemexpress LLC HY-136485 5mg Medchemexpress, FEN1-IN-4 CAS:1995893-58-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364855MG
Encompass_Preferred
Medchemexpress, HY-136485 5mg FEN1-IN-4 CAS:1995893-58-7 FEN1-IN-4 (Compound 2) is a human flap endonuclease-1 (hFEN1) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0446