Learn More
Cayman Chemical Fenbendazole 100g
SDP
Supplier:  Cayman Chemical 19687100 G
Encompass_Preferred
A benzimidazole anthelmintic; active against Giardia in vitro (IC50 = 0.3 μM); prevents infiltration of parasites into the brain in a rabbit model of E. cuniculi infection at 20 mg/kg; activates HIF-1α and prevents oxidative stress-induced death in primary neurons in vitro
Catalog No. 50-298-3370