Learn More
Selleck Chemical LLC Fenbendazole S2468-50mg
SDP
Supplier:  Selleck Chemical LLC S246850MG
Encompass_Preferred
Fenbendazole(Fenbendazol Phenbendasol) is a broad spectrum benzimidazole anthelmintic used against gastrointestinal parasites with an IC50 of about 0 01 g/mL
Catalog No. 50-311-2391