Learn More
Cayman Chemical Fenbendazole-d3 5mg
SDP
Supplier:  Cayman Chemical 278425 MG
Encompass_Preferred
An internal standard for the quantification of fenbendazole by GC- or LC-MS
Catalog No. 50-294-0057