Learn More
Cayman Chemical Fenbendazole sulfon 10mg
SDP
Supplier:  Cayman Chemical 2092110 MG
Encompass_Preferred
A minor metabolite of fenbendazole; formed by oxidation of oxfendazole, a product of fenbendazole oxidation
Catalog No. 50-298-1624