Learn More
Medchemexpress LLC (-)-Fenchone | 7787-20-4 | 1 G

Supplier: Medchemexpress LLC HYN51321G
(-)-Fenchone is a chemical compound with the molecular formula C10H16O and a molecular weight of 152.24. It is typically a colorless to light yellow liquid. This product is provided as a standard, with a purity of 98.0% by NMR, and is intended for research use only. It has not been fully validated for medical applications.
- CAS number: 7787-20-4
- Molecular formula: C10H16O
- Molecular weight: 152.24
- Appearance: Colorless to light yellow liquid
- Purity: 98.0% (NMR)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.