Learn More
Medchemexpress LLC Fenclozic acid | 17969-20-9 | MFCD00721754 | 99.5% | 253.70 | C11H8ClNO2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1014325MG
Encompass_Preferred
Fenclozic acid is a small-molecule research compound with analgesic, antipyretic, and anti-inflammatory activity. It is supplied for laboratory use as a solid powder and as a DMSO solution. Chemical identity: C11H8ClNO2S; CAS 17969-20-9; mol. wt. 253.70.
  • Orally active analgesic, antipyretic, and anti-inflammatory agent.
  • High purity suitable for research applications.
  • Available in small milligram packages and solution formats.
  • Provided with analytical data including COA and SDS.
  • Stable under recommended storage conditions when handled per SDS.
Catalog No. 50-002-47034