Learn More
Medchemexpress LLC Fendiline hydrochloride | 13636-18-5
SDP
Supplier:  Medchemexpress LLC 250MGHYB0984
Encompass_Preferred
Fendiline hydrochloride is a diphenylalkylamine type of antianginal agent that functions as an L-type calcium channel blocker. It also acts as a selective K-Ras inhibitor, without affecting H-Ras and N-Ras. Furthermore, it is identified as a STING agonist and has demonstrated the ability to inhibit the growth of various refractory cold tumors.
  • Functions as an L-type calcium channel blocker (IC50 of 17 μM)
  • Acts as a selective K-Ras inhibitor, without affecting H-Ras and N-Ras
  • Inhibits K-Ras plasma membrane localization (IC50 of 9.64 μM)
  • Blocks proliferation of pancreatic, colon, lung, and endometrial cancer cell lines expressing oncogenic mutant K-Ras
  • Identified as a STING agonist
  • Inhibits growth of multiple refractory cold tumors (MC38, CT26, and B16F10)
Catalog No. 50-004-05625