Learn More
Medchemexpress LLC Fendiline hydrochloride | 13636-18-5 | 99.8% | 351.91 | 100 MG

Supplier: Medchemexpress LLC 100MGHYB0984
Fendiline hydrochloride is a diphenylalkylamine type of antianginal agent that functions as an L-type calcium channel blocker and selective K-Ras inhibitor. It inhibits K-Ras plasma membrane localization, thereby blocking K-Ras signal output and the proliferation of various cancer cell lines. Additionally, it acts as a STING agonist, capable of inhibiting the growth of multiple refractory cold tumors.
- Functions as an L-type calcium channel blocker
- Acts as a selective K-Ras inhibitor
- Inhibits K-Ras plasma membrane localization
- Blocks proliferation of pancreatic, colon, lung, and endometrial cancer cell lines
- Acts as a STING agonist capable of inhibiting tumor growth
- Purity: 99.78%
- Molecular weight: 351.91
- Formula: C23H26ClN
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.