Learn More
Selleck Chemical LLC Fenebrutinib
SDP
Supplier:  Selleck Chemical LLC S84215MG
Encompass_Preferred
Fenebrutinib (GDC-0853) is a potent selective and non-covalent bruton s tyrosine kinase (BTK) inhibitor with an Ki value of 0 91 nM for Btk with 100-fold selectivity over 3 off-targets (Bmx 153-fold Fgr 168-fold Src 131-fold)
Catalog No. 50-000-00014