Learn More
Roboz Surgical 3.5'' Long Fenestrated Forceps
SDP
Supplier:  Roboz Surgical RS5161
Encompass
3.5" long fenestrated forceps. The external diameter of the round fenestrated tip is 3 mm. The internal diameter of the fenestration is 2.2 mm. The tips are not serrated.
Catalog No. NC1742182