Learn More
Medchemexpress LLC Fenhexamid | 126833-17-8 | MFCD03095700 | 99.6% | 302.2 g/mol | C14H17Cl2NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118065100MG
Encompass_Preferred
Fenhexamid is a hydroxyanilide fungicide that inhibits sterol biosynthesis in fungal pathogens and is active against Botrytis cinerea. Supplied as a solid research reagent, it is used for antifungal activity studies and analytical applications.
  • Acts as a sterol biosynthesis inhibitor effective against Botrytis cinerea.
  • High purity suitable for research and analytical use (99.6%).
  • Chemical formula C14H17Cl2NO2; molecular weight 302.2 g/mol.
  • Provided as a solid in small research quantities, including 100 mg sizes.
  • Commonly used in fungicide efficacy and resistance research.
Catalog No. 50-002-26505